L(+)-Arabinose, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5328-37-0
:87-72-9
Formula:C₅H₁₀O₅
MDL Number:MFCD00067709
:150.13 g/mol
Boiling Pt:331 °C (1013 hPa)
Melting Pt:158…160 °C
Density:1.585 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:226-214-6