Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Hydroxy-3,4-dimethoxybenzaldehyde
5-Hydroxy-3,4-dimethoxybenzaldehyde, Sigma-Aldrich®
5-Hydroxy-3,4-dimethoxybenzaldehyde, Sigma-Aldrich®
5-Hydroxy-3,4-dimethoxybenzaldehyde, Sigma-Aldrich®
SIAL258717-1G
:
Merck
:
5-Hydroxy-3,4-dimethoxybenzaldehyde, Sigma-Aldrich®
SIAL258717-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
M-Clarity™ quality level = MQ100
:
182.1733 g/mol
:
29865-90-5
:
C₉H₁₀O₄
:
MFCD00010442