
3,5-Dichloro-2-hydroxybenzenesulphonyl chloride ≥97%
Catalog # L12147.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:261.51 g/mol
Melting Pt:78…80 °C
Storage Temperature:Ambient
EINECS:245-619-9
UN:3261
ADR:8,III
:GHS05
CAS Number:23378-88-3
Formula:C₆H₃Cl₃O₃S
MDL Number:MFCD00007432