Order Entry
Export
ContactUsLinkComponent
Ammonium chloride-[¹⁵N] (60-80% ¹⁵N), Sigma-Aldrich®
Ammonium chloride-[¹⁵N] (60-80% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL488003-5G
Supplier:  Merck
CAS Number:  
Ammonium chloride-[¹⁵N] (60-80% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL488003-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:39466-62-1
Formula:H₄ClN
MDL Number:MFCD00075417