Order Entry
Export
Orders LinkContactUsLinkComponent
Ammonium chloride-[¹⁵N] (98% ¹⁵N)

Ammonium chloride-[¹⁵N] (98% ¹⁵N)

Supplier:   Merck
Ammonium chloride-[¹⁵N] (98% ¹⁵N)
Supplier:  Merck
Certificates
Ammonium chloride-[¹⁵N] (98% ¹⁵N)

5 Options Available Below

Product Details & Documents
Synonyms:  Ammonium-15N chloride, Ammonium chloride-15N
  • CAS Number:39466-62-1
  • Formula:H₄ClN
  • MDL Number:MFCD00075417
  •  :M-Clarity™ quality level = MQ200
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 125,00 €
    125,00 €
    1 * 250 mg
       125,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 138,00 €
    138,00 €
    1 * 1 g
       138,00 €

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 1 010,00 €
    1 010,00 €
    1 * 10 g
       1 010,00 €

  • Glass bottle for solids
    20 g
    Log in for availability
    Log in for availability
    1 * 20 g
    1 * 20 g - 1 790,00 €
    1 790,00 €
    1 * 20 g
       1 790,00 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 3 970,00 €
    3 970,00 €
    1 * 50 g
       3 970,00 €