Order Entry
Export
Orders LinkContactUsLinkComponent
 

Hydrocortisone-21-acetate

Glass bottle for solids, 100 mg
Catalog # PROOBP584
Hydrocortisone-21-acetate
Catalog # PROOBP584
CAS Number:  50-03-3
Hydrocortisone-21-acetate

Glass bottle for solids, 100 mg

CAS Number:  50-03-3
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Hydrocortisone acetate
  • CAS Number:50-03-3
  • Formula:C₂₃H₃₂O₆
  • MDL Number:MFCD00037714
  • Molecular Weight:404.51 g/mol
  • EINECS:200-004-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications