Order Entry
Export
ContactUsLinkComponent
 
Organic reference standard, Estradiol 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA13213100AL
 :  
Organic reference standard, Estradiol 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA13213100AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    50-28-2
  • Description:
    Estradiol 100 µg/ml in acetonitrile
  • Formula:
    C₁₈H₂₄O₂
  • Description:
    17- -Estradiol hemihydrate