4'-Hydroxy-3'-methylacetophenone
Catalog # MAYBSB01922.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:150.18 g/mol
EINECS:212-880-5
CAS Number:876-02-8
Formula:C₉H₁₀O₂
MDL Number:MFCD00002231