Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Iodo-3,4-dihydro-2H-1,5-benzodioxepine
7-Iodo-3,4-dihydro-2H-1,5-benzodioxepine
7-Iodo-3,4-dihydro-2H-1,5-benzodioxepine
7-Iodo-3,4-dihydro-2H-1,5-benzodioxepine
Supplier:
Thermo Scientific
Certificates
Expand image
7-Iodo-3,4-dihydro-2H-1,5-benzodioxepine
Supplier:
Thermo Scientific
About this item
CAS Number:
306934-90-7
:
Formula:
C₉H₉IO₂
MDL Number:
MFCD02677706