Order Entry
Export
ContactUsLinkComponent
 
Pesticide reference standard, Disulfoton 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA12980000CY
 :  
Pesticide reference standard, Disulfoton 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA12980000CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    298-04-4
  • Description:
    Disulfoton 100 µg/ml in cyclohexane
  • Formula:
    C₈H₁₉O₂PS₃
  • Description:
    Disulfoton