Pesticide reference standard, Disulfoton 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA12980000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:298-04-4
- Description:Disulfoton 100 µg/ml in acetonitrile
- Formula:C₈H₁₉O₂PS₃
- Description:Disulfoton