Order Entry
Export
ContactUsLinkComponent
 
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene, Sigma-Aldrich®
Catalog # SIALD65607-50G
Supplier:  Merck
CAS Number:  
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene, Sigma-Aldrich®
Catalog # SIALD65607-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :217.14 g/mol
ADR:8,III
 :M-Clarity™ quality level = MQ100
UN:3261
CAS Number:1585-17-7
Formula:C₁₁H₁₄Cl₂
MDL Number:MFCD00013681