Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene
Supplier:
Thermo Scientific
Certificates
Expand image
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene
Supplier:
Thermo Scientific
About this item
CAS Number:
1585-17-7
:
Formula:
C₁₁H₁₄Cl₂
MDL Number:
MFCD00013681
:
217.14 g/mol
UN:
3261
ADR:
8,III