
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1585-17-7
:
Formula:C₁₁H₁₄Cl₂
MDL Number:MFCD00013681
:217.14 g/mol
Boiling Pt:180 °C (20 mmHg)
Melting Pt:104…107 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05