Order Entry
Export
Orders LinkContactUsLinkComponent
 

Erucic acid

Erucic acid
Certificates

4 Options Available Below

 
Synonyms:  (Z)-Docos-13-enoic acid
  • CAS Number:112-86-7
  • Formula:C₂₂H₄₂O₂
  • MDL Number:MFCD00063188
  • Molecular Weight:338.58 g/mol
  • EINECS:204-011-3
Product Family Options

Product Information

  • Pack type
    Pk
     
     
  • Glass bottle for solids
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 15,00 €
    15,00 €
    1 * 50 mg
       15,00 €

  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 24,00 €
    24,00 €
    1 * 100 mg
       24,00 €

  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 53,00 €
    53,00 €
    1 * 250 mg
       53,00 €

  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 93,00 €
    93,00 €
    1 * 500 mg
       93,00 €