3-Acetylbenzophenone
Glass bottle for solids, 100 mg
Catalog # PROOMM0001.01
Supplier: LGC Standards PROMOCHEM
3-Acetylbenzophenone
Catalog # PROOMM0001.01
Supplier: LGC Standards PROMOCHEM
CAS Number: 66067-44-5
Jump to:
Product Details & Documents
Synonyms: 1-(3-Benzoylphenyl)ethan-1-one, 1-(3-Benzoylphenyl)ethanone
- CAS Number:66067-44-5
- Formula:C₁₅H₁₂O₂
- MDL Number:MFCD01662280
- Molecular Weight:224.2546 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



