2,5-Dichloronicotinamide
Supplier: Thermo Scientific
Jump to:
About this item
- CAS Number:75291-86-0
- Formula:C₆H₄Cl₂N₂O
- MDL Number:MFCD00204159
2 Options Available Below
Product Details & Documents
- CAS Number:75291-86-0
- Formula:C₆H₄Cl₂N₂O
- MDL Number:MFCD00204159
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



