Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Cellobiose octaacetate

D-(+)-Cellobiose octaacetate

Glass bottle for solids, 100 g
Catalog # MOLEM82788502
Supplier:   Molekula
D-(+)-Cellobiose octaacetate
Catalog # MOLEM82788502
Supplier:  Molekula
CAS Number:  5346-90-7
D-(+)-Cellobiose octaacetate

Glass bottle for solids, 100 g

CAS Number:  5346-90-7
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:5346-90-7
  • Formula:C₂₈H₃₈O₁₉
  • MDL Number:MFCD00009600
  • Molecular Weight:678.59 g/mol
  • Storage Temperature:Ambient
  • EINECS:226-304-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications