D-(+)-Cellobiose octaacetate
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 5346-90-7 Molecular Formula: C28H38O19 Formula Weight: 678.59 Melting Point: 224-231° Beilstein Reference: 79280 Merck Reference: 14,1961 Optical Rotation: +40° (c=1 in chloroform) MDL No.: MFCD00009600
- CAS Number:5346-90-7
- Formula:C₂₈H₃₈O₁₉
- MDL Number:MFCD00009600
- Molecular Weight:678.59 g/mol
- Merck Index:14,1961
- Melting Pt:224…231 °C
- Storage Temperature:Ambient
- EINECS:226-304-5
2 Options Available Below
Product Details & Documents
CAS No.: 5346-90-7 Molecular Formula: C28H38O19 Formula Weight: 678.59 Melting Point: 224-231° Beilstein Reference: 79280 Merck Reference: 14,1961 Optical Rotation: +40° (c=1 in chloroform) MDL No.: MFCD00009600
- CAS Number:5346-90-7
- Formula:C₂₈H₃₈O₁₉
- MDL Number:MFCD00009600
- Molecular Weight:678.59 g/mol
- Merck Index:14,1961
- Melting Pt:224…231 °C
- Storage Temperature:Ambient
- EINECS:226-304-5



