Order Entry
Export
Orders LinkContactUsLinkComponent
trans,trans-Dibenzylideneacetone 98%

trans,trans-Dibenzylideneacetone 98%

trans,trans-Dibenzylideneacetone 98%
Certificates
trans,trans-Dibenzylideneacetone 98%

3 Options Available Below

Product Details & Documents
Synonyms:  (1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one
  • CAS Number:35225-79-7
  • Formula:C₁₇H₁₄O
  • MDL Number:MFCD00004790
  • Molecular Weight:234.30 g/mol
  • Melting Pt:107…114 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    108 to 114 °C
  • Assay:
    ≥97.5 %
  • Pore volume:
    0.80 to 1.00 ml/g
  • Carbon (C):
    ≥16 %
  • Volatile matter:
    ≤4 % (110 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 31,10 €
    31,10 €
    1 * 5 g
       31,10 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 92,80 €
    92,80 €
    1 * 25 g
       92,80 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 221,00 €
    221,00 €
    1 * 100 g
       221,00 €