Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III), Sigma-Aldrich®
Catalog # SIAL547778-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:620.06 g/mol
ADR:4.3,II
:M-Clarity™ quality level = MQ100
UN:3396
CAS Number:175923-07-6
:35788-99-9
Formula:C₁₈H₅₄LaN₃Si₆
MDL Number:MFCD00271009