Order Entry
Export
Orders LinkContactUsLinkComponent
4-Bromo-3,5-dimethyl-1H-pyrazole

4-Bromo-3,5-dimethyl-1H-pyrazole

Supplier:   Thermo Scientific
4-Bromo-3,5-dimethyl-1H-pyrazole
Supplier:  Thermo Scientific
Certificates
4-Bromo-3,5-dimethyl-1H-pyrazole

2 Options Available Below

Product Details & Documents
Synonyms:  4-Bromo-3,5-dimethylpyrazole
  • CAS Number:3398-16-1
  • Formula:C₅H₇BrN₂
  • MDL Number:MFCD00005242
  • Molecular Weight:175.03 g/mol
  • EINECS:222-263-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g