Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dicyano-2-phenylimidazole
4,5-Dicyano-2-phenylimidazole ≥97%
4,5-Dicyano-2-phenylimidazole ≥97%
H58800.04
:
Thermo Fisher Scientific
:
4,5-Dicyano-2-phenylimidazole ≥97%
H58800.04
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
2 g
:
:
194.19 g/mol
:
Ambient
:
3439
:
6.1,III
:
GHS07
:
50847-06-8
:
C₁₁H₆N₄
:
MFCD05863325