Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Trimethylsilyl)ethoxymethyl chloride
2-(Trimethylsilyl)ethoxymethyl chloride, Sigma-Aldrich®
2-(Trimethylsilyl)ethoxymethyl chloride, Sigma-Aldrich®
2-(Trimethylsilyl)ethoxymethyl chloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2-(Trimethylsilyl)ethoxymethyl chloride, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
76513-69-4
:
Formula:
C₆H₁₅ClOSi
MDL Number:
MFCD00009919
:
166.72 g/mol
EINECS:
278-483-4
UN:
2920
ADR:
8,II