
3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:175136-66-0
:
Formula:C₁₇H₆F₁₂O
MDL Number:MFCD00042474
:454.22 g/mol
Melting Pt:139…140 °C
:GHS07
Storage Temperature:Ambient