3,4-Dimethylbenzophenone ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 2571-39-3 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 44-47° Boiling Point: 335° Beilstein Reference: 1952043 MDL No.: MFCD00008525
Synonyms: (3,4-Dimethylphenyl)(phenyl)methanone
- CAS Number:2571-39-3
- Formula:C₁₅H₁₄O
- MDL Number:MFCD00008525
- Molecular Weight:210.28 g/mol
- Boiling Pt:335 °C
- Melting Pt:44…47 °C
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:219-916-9
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 2571-39-3 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 44-47° Boiling Point: 335° Beilstein Reference: 1952043 MDL No.: MFCD00008525
Synonyms: (3,4-Dimethylphenyl)(phenyl)methanone
- CAS Number:2571-39-3
- Formula:C₁₅H₁₄O
- MDL Number:MFCD00008525
- Molecular Weight:210.28 g/mol
- Boiling Pt:335 °C
- Melting Pt:44…47 °C
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:219-916-9



