
7-(2,3-Dihydroxypropyl)theophylline 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:479-18-5
:
Formula:C₁₀H₁₄N₄O₄
MDL Number:MFCD00005759
:254.24 g/mol
Melting Pt:160…165 °C
EINECS:207-526-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:160 to 165 °C
- Loss on drying:≤0.5 % (1 g, 105 °C)
- Titration with HClO4:≥98.5 % (On dried substance)