Order Entry
Export
Orders LinkContactUsLinkComponent
Bis(pinacolato)diboron ≥98%

Bis(pinacolato)diboron ≥98%

Bis(pinacolato)diboron ≥98%
Certificates
Bis(pinacolato)diboron ≥98%
Bis(pinacolato)diboron ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 73183-34-3 Molecular Formula: C12H24B2O4 Formula Weight: 253.94 Storage Temperature: 2°C to 8°C Melting Point: 133-139° Beilstein Reference: 7703552 MDL No.: MFCD00799570

Synonyms:  4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
  • CAS Number:73183-34-3
  • Formula:C₁₂H₂₄B₂O₄
  • MDL Number:MFCD00799570
  • Molecular Weight:253.94 g/mol
  • Melting Pt:133…139 °C
Product Family Options

Product Information