Order Entry
Export
Orders LinkContactUsLinkComponent
Folic acid

Folic acid

Glass bottle for solids, 100 mg
Catalog # PROOICRS0316
Folic acid
Catalog # PROOICRS0316
CAS Number:  59-30-3
Folic acid

Glass bottle for solids, 100 mg

CAS Number:  59-30-3
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
  • CAS Number:59-30-3
  • Formula:C₁₉H₁₉N₇O₆
  • MDL Number:MFCD00079305
  • Molecular Weight:441.41 g/mol
  • EINECS:200-419-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications