[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:95464-05-4
:
Formula:C₃₅H₃₀Cl₄FeP₂Pd
MDL Number:MFCD00792899
:816.64 g/mol
:M-Clarity™ quality level = MQ200