Order Entry
Export
ContactUsLinkComponent
4'-Amino-2'-hydroxyacetophenone
4'-Amino-2'-hydroxyacetophenone
Catalog # CHMPBTB10056.250
Supplier:  CHEMPUR
CAS Number:  
4'-Amino-2'-hydroxyacetophenone
Catalog # CHMPBTB10056.250
Supplier:  CHEMPUR
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :151.1600 g/mol
CAS Number:2476-29-1
Formula:C₈H₉NO₂
MDL Number:MFCD00100636