Specifications
- Description:Precast DIGE gel
- Environmentally Preferable:
- Pk:3
Specifications
About this item
DIGE gel is a 12,5% precast polyacrylamide gel in a low fluorescent glass cassette for second dimension 2-D electrophoresis and specially developed for 2-D DIGE analysis. DIGE gel and DIGE buffer kit are tools to increase the quality, reproducibility, and convenience of 2-D DIGE using Ettan™ DIGE system.
- Ready to use products with a long shelf-life that saves you time by eliminating the need to make buffers, cast gels, and wash glass plates
- Provides high sensitivity and high resolution protein spot maps of equal quality compared to homecast polyacrylamide gels
- Runs with DIGE buffer kit result in spot maps with higher reproducibility compared to homecast polyacrylamide gels
Delivery: Please be aware that this product may be shipped 30 days before the expiry date. For more information about batch specific expiry dates please contact Avantor technical service.