Laboratory trays
Catalog # VITL165094
Supplier: VITLAB
Specifications
- Colour:White
- Seq:01
- W×D×H:230×180x42 mm
- Pk:1
Specifications
About this item
Laboratory trays, made of white PP.
- Robust design
- Easy to clean
- Good chemical resistance
- Smooth surface, rounded corners and edges
Ideal for a wide range of applications.