Order Entry
Export
ContactUsLinkComponent
Gel filtration media, Sephacryl™ High Resolution
Gel filtration media, Sephacryl™ High Resolution
Supplier:  Cytiva
Certificates
undefined
Gel filtration media, Sephacryl™ High Resolution
Supplier:  Cytiva
Certificates

Specifications

  • pH range:
    3 - 11
  • Matrix:
    Cross-linked copolymer of allyl dextran and N,N'-methylene bisacrylamide
  • Storage temperature:
    4 - 30 °C

Specifications