Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Arsenazo III
Arsenazo III
Arsenazo III
Toxic
Environment
Arsenazo III
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Toxic
Environment
Arsenazo III
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
1668-00-4
:
Formula:
C₂₂H₁₈As₂N₄O₁₄S₂
MDL Number:
MFCD00036695
:
776.38 g/mol
Storage Temperature:
Ambient
EINECS:
216-788-6
UN:
3465
ADR:
6.1,III
:
GHS06
:
GHS09
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ63031
Videos
Thermo Scientific Acroseal 25 Years
Documentation