Melatonin ≥99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 73-31-4 Molecular Formula: C13H16N2O2 Formula Weight: 232.28 Storage Temperature: 2°C to 8°C Physical Form: Powder Melting Point: 116-120° Solubility: Soluble in ethanol to at least 50mg/ml Beilstein Reference: 205542 Merck Reference: 14,5816 MDL No.: MFCD00005655
A pineal gland hormone associated with sleep regulation. It induces apoptosis
Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
- CAS Number:73-31-4
- Formula:C₁₃H₁₆N₂O₂
- MDL Number:MFCD00005655
- Molecular Weight:232.28 g/mol
- Merck Index:14,5816
- Melting Pt:116…120 °C
- EINECS:200-797-7
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Cell Culture Workflow
Product Literature
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



