Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dexamethasone 21-acetate
Dexamethasone-21-acetate
Dexamethasone-21-acetate
Irritant
Dexamethasone-21-acetate
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Dexamethasone-21-acetate
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
1177-87-3
:
Formula:
C₂₄H₃₁FO₆
MDL Number:
MFCD00027407
:
434.5 g/mol
Merck Index:
14,2943
Melting Pt:
240 °C
EINECS:
214-646-8
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ63691
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation