
Guanidinium hydrochloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50-01-1
:
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026
:95.53 g/mol
Merck Index:14,4562
Melting Pt:185…188 °C
Density:1.344
EINECS:200-002-3
:GHS07
Storage Temperature:Ambient