Benzamide
Catalog # EHERC10532250
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:121.14 g/mol
Merck Index:13,01059
Boiling Pt:288 °C (1013 hPa)
Melting Pt:124 °C
Density:1.35 g/cm³ (20 °C)
Flash Pt:180 °C
EINECS:200-227-7
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:55-21-0
Formula:C₇H₇NO
MDL Number:MFCD00007968