Order Entry
Export
ContactUsLinkComponent
2,4-Dichloro-L-phenylalanine ≥98%
2,4-Dichloro-L-phenylalanine ≥98%
Catalog # H27579.03
CAS Number:  
2,4-Dichloro-L-phenylalanine ≥98%
Catalog # H27579.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:111119-36-9
 :
Formula:C₉H₉Cl₂NO₂
MDL Number:MFCD01860882
 :234.1 g/mol
Storage Temperature:Ambient