Order Entry
Export
ContactUsLinkComponent
3-(3,4-Methylenedioxyphenyl)propionic acid ≥97%, Sigma-Aldrich®
Irritant
3-(3,4-Methylenedioxyphenyl)propionic acid ≥97%, Sigma-Aldrich®
Catalog # SIAL657565-1G
Supplier:  Merck
CAS Number:  
Irritant
3-(3,4-Methylenedioxyphenyl)propionic acid ≥97%, Sigma-Aldrich®
Catalog # SIAL657565-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.19 g/mol
Melting Pt:83…87 °C
 :M-Clarity™ quality level = MQ100
EINECS:220-565-9
 :GHS07
CAS Number:2815-95-4
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00016909