Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydrogen chloride
Hydrogen chloride 1 M in acetic acid
Hydrogen chloride 1 M in acetic acid
:
Thermo Fisher Scientific
Hydrogen chloride 1 M in acetic acid
:
Thermo Fisher Scientific
:
7647-01-0
:
:
HCl
:
MFCD00011324
:
36.46 g/mol
:
1.050
:
40 °C (104 °F)
:
231-595-7
:
2920
:
8,II
:
GHS05
:
GHS02
:
GHS07
:
Ambient
msds_ALFAH27347
Thermo Scientific Acroseal 25 Years