3,5-Dimethoxybenzyl bromide, Sigma-Aldrich®
Catalog # SIAL480622-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:231.0864 g/mol
CAS Number:877-88-3
Formula:C₉H₁₁BrO₂
MDL Number:MFCD01321368