Silicon nitride
Catalog # PROONIST-1900
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:4 g
- Seq:3155
Specifications
About this item
:
:140.28 g/mol
EINECS:234-796-8
CAS Number:12033-89-5
Formula:N₄Si₃
MDL Number:MFCD00011230