
4-(Aminomethyl)phenylboronic acid hydrochloride 96%
Catalog # ACRO358842500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:Light yellow Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥95 %
Specification Test Results
About this item
:
:187.43 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:75705-21-4
Formula:C₇H₁₁BClNO₂
MDL Number:MFCD01632199