

Hexafluorophosphoric acid 60% (w/w) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16940-81-1
:
Formula:HF₆P
MDL Number:MFCD00011350
:145.97 g/mol
Density:1.75 g/cm³
Storage Temperature:Ambient
EINECS:241-006-5
UN:2922
ADR:8,II
:GHS06
:GHS05
Specification Test Results
- Appearance:Colorless to yellow-green Solution
- Assay Titrimetry:≥55.0 %
- Additional info:assay based on PF6 anion-content