Order Entry
Export
ContactUsLinkComponent
Acetic anhydride-[2,2'-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Acetic anhydride-[2,2'-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL487848-250MG
Supplier:  Merck
CAS Number:  
Acetic anhydride-[2,2'-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL487848-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
ADR:8,II
 :M-Clarity™ quality level = MQ200
UN:1715
CAS Number:98006-45-2
Formula:C₄H₆O₃
MDL Number:MFCD00083863