Order Entry
Export
ContactUsLinkComponent
Acetic anhydride-[1,1'-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Acetic anhydride-[1,1'-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL487813-500MG
Supplier:  Merck
CAS Number:  
Acetic anhydride-[1,1'-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL487813-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:90980-78-2
Formula:C₄H₆O₃
MDL Number:MFCD00083862