2,2'-Dibenzothiazole disulphide, Sigma-Aldrich®
Catalog # SIALD218154-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:332.4867 g/mol
:M-Clarity™ quality level = MQ100
EINECS:204-424-9
CAS Number:120-78-5
Formula:C₁₄H₈N₂S₄
MDL Number:MFCD00022874