Order Entry
Export
ContactUsLinkComponent
1,4-Thioxane, Sigma-Aldrich®
1,4-Thioxane, Sigma-Aldrich®
Catalog # SIAL131970-25G
Supplier:  Merck
CAS Number:  
1,4-Thioxane, Sigma-Aldrich®
Catalog # SIAL131970-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :104.17 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:240-117-6
CAS Number:15980-15-1
Formula:C₄H₈OS
MDL Number:MFCD00006573